Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnd1
Entrez Gene ID
733500
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
Xdead, dead-end, dnd, rbms4, xde
Chromosome Location
Chr.3: 17722384 - 17736262 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044434.1 NP_001037899.1 3 - 4 676 93
GenBank mRNA CR760562.2 - 3 - 4 676 93

View Full Product Details