Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc38a8
Entrez Gene ID
733502
Gene Name
solute carrier family 38 member 8
Gene Aliases
snat8
Chromosome Location
Chr.4: 50826428 - 50851978 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044435.1 NP_001037900.1 9 - 10 1478 77
XM_012961231.2 XP_012816685.1 10 - 11 1279 77
XM_012961232.2 XP_012816686.1 10 - 11 1328 77
XM_018093079.1 XP_017948568.1 10 - 11 1291 77
GenBank mRNA CR760593.2 - 9 - 10 1478 77

View Full Product Details