Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
thdl18
Entrez Gene ID
733520
Gene Name
thyroid hormone down-regulated protein (gene 18)
Gene Aliases
gene18
Chromosome Location
Chr.2: 50989752 - 51002888 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044446.1 NP_001037911.1 7 - 8 1060 74
XM_018091315.1 XP_017946804.1 5 - 6 917 74
GenBank mRNA CR760814.2 - 7 - 8 1060 74

View Full Product Details