Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mbd4
Entrez Gene ID
733528
Gene Name
methyl-CpG binding domain 4, DNA glycosylase
Gene Aliases
med1
Chromosome Location
Chr.4: 121414433 - 121424886 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044451.1 NP_001037916.1 3 - 4 893 92
GenBank mRNA CR760952.1 - 3 - 4 893 92

View Full Product Details