Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ociad2
Entrez Gene ID
733710
Gene Name
OCIA domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 30007293 - 30014996 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044487.1 NP_001037952.1 4 - 5 350 70
GenBank mRNA BC076708.1 - 4 - 5 221 70
BC155045.1 - 4 - 5 322 70
BC155046.1 - 4 - 5 322 70
CR760011.2 - 4 - 5 350 70
CR761639.2 - 4 - 5 331 70

View Full Product Details