Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nptn
Entrez Gene ID
733721
Gene Name
neuroplastin
Gene Aliases
-
Chromosome Location
Chr.3: 82826339 - 82866974 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044493.1 NP_001037958.1 1 - 2 136 94
XM_012966685.2 XP_012822139.1 1 - 2 325 94
GenBank mRNA BC170738.1 - 1 - 2 122 94
BC170742.1 - 1 - 2 122 94
CR760664.2 - 1 - 2 136 94

View Full Product Details