Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c8orf76
Entrez Gene ID
733825
Gene Name
chromosome 8 open reading frame 76
Gene Aliases
-
Chromosome Location
Chr.7: 4855112 - 4862175 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045584.1 NP_001039049.1 4 - 5 880 85
GenBank mRNA BC161270.1 - 4 - 5 807 85
CR848448.2 - 4 - 5 880 85

View Full Product Details