Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsf3
Entrez Gene ID
733835
Gene Name
heat shock transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.8: 19631221 - 19651120 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045588.1 NP_001039053.1 6 - 7 605 86
GenBank mRNA CR848515.2 - 6 - 7 605 86

View Full Product Details