Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
fxyd3
Entrez Gene ID
733884
Gene Name
FXYD domain-containing ion transport regulator 3
Gene Aliases
-
Chromosome Location
Chr.7: 117135929 - 117160954 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045616.1 NP_001039081.1 - 111 62
GenBank mRNA CR926162.2 - - 111 62

View Full Product Details