Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gzf1
Entrez Gene ID
733920
Gene Name
GDNF inducible zinc finger protein 1
Gene Aliases
zbtb23, znf336
Chromosome Location
Chr.5: 20791942 - 20801440 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045636.1 NP_001039101.1 3 - 4 1637 91
XM_012963100.1 XP_012818554.1 4 - 5 1670 91
XM_012963101.2 XP_012818555.1 4 - 5 1731 91
XM_012963102.2 XP_012818556.1 4 - 5 1736 91
GenBank mRNA BC121586.1 - 3 - 4 1179 91
BC171267.1 - 3 - 4 1630 91
CR942387.2 - 3 - 4 1637 91

View Full Product Details