The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045658.1 | NP_001039123.1 | 23 - 24 | 2288 | 103 |
XM_012964360.1 | XP_012819814.1 | 23 - 24 | 2288 | 103 | |
XM_012964366.2 | XP_012819820.1 | 23 - 24 | 2288 | 103 | |
GenBank mRNA | BC168434.1 | - | 23 - 24 | 2202 | 103 |
CR942469.2 | - | 23 - 24 | 2288 | 103 |