Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
utp23
Entrez Gene ID
733957
Gene Name
UTP23, small subunit processome component
Gene Aliases
-
Chromosome Location
Chr.6: 121572378 - 121575670 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045669.1 NP_001039134.1 1 - 2 230 72
GenBank mRNA CR942499.2 - 1 - 2 230 72

View Full Product Details