Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prdm8
Entrez Gene ID
733976
Gene Name
PR domain containing 8
Gene Aliases
-
Chromosome Location
Chr.1: 76500661 - 76505528 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045686.1 NP_001039151.1 3 - 4 722 73
GenBank mRNA BC136202.1 - 3 - 4 670 73
CR942568.2 - 3 - 4 722 73

View Full Product Details