Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psmc1
Entrez Gene ID
733980
Gene Name
proteasome 26S subunit, ATPase 1
Gene Aliases
p26s4, p56, pros26.4
Chromosome Location
Chr.8: 65646815 - 65656451 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045689.2 NP_001039154.1 5 - 6 539 96
GenBank mRNA CR942586.2 - 4 - 5 468 96
CX433740.2 - 5 - 6 537 96

View Full Product Details