Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lonrf2
Entrez Gene ID
733984
Gene Name
LON peptidase N-terminal domain and ring finger 2
Gene Aliases
-
Chromosome Location
Chr.2: 108737620 - 108788172 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045693.1 NP_001039158.1 4 - 5 1417 70
XM_012957170.2 XP_012812624.1 4 - 5 1484 70
GenBank mRNA BC121307.1 - 4 - 5 1417 70
CR942594.2 - 4 - 5 1332 70

View Full Product Details