Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stoml1
Entrez Gene ID
734047
Gene Name
stomatin like 1
Gene Aliases
-
Chromosome Location
Chr.3: 83133465 - 83149646 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045728.1 NP_001039193.1 5 - 6 963 94
XM_018092141.1 XP_017947630.1 5 - 6 1135 94
GenBank mRNA CR926277.2 - 5 - 6 963 94

View Full Product Details