Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mcm3
Entrez Gene ID
734092
Gene Name
minichromosome maintenance complex component 3
Gene Aliases
zmcm3
Chromosome Location
Chr.Un|NW_016683369: 986871 - 999596 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045766.1 NP_001039231.1 14 - 15 2133 74
GenBank mRNA BC167938.1 - 14 - 15 2124 74
CR942676.2 - 14 - 15 2133 74

View Full Product Details