Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nme4
Entrez Gene ID
734101
Gene Name
NME/NM23 nucleoside diphosphate kinase 4
Gene Aliases
NM23-X4, ndpk-d, nm23-h4, nm23h4, nme4-a, nme4-b
Chromosome Location
Chr.9: 15194270 - 15202354 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045774.1 NP_001039239.1 3 - 4 343 88
XM_012970515.2 XP_012825969.1 3 - 4 411 88
GenBank mRNA BC158417.1 - 3 - 4 373 88
CR942753.2 - 3 - 4 343 88

View Full Product Details