Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC548369
Entrez Gene ID
548369
Gene Name
uncharacterized LOC548369
Gene Aliases
-
Chromosome Location
Chr.1: 128398510 - 128500803 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC087793.1 - 5 - 6 732 84

View Full Product Details