Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
eln1
Entrez Gene ID
779443
Gene Name
tropoelastin 1
Gene Aliases
eln
Chromosome Location
Chr.2: 37685140 - 37763602 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078709.1 NP_001072177.1 20 - 21 1331 78
XM_012957185.2 XP_012812639.1 22 - 23 1517 78
GenBank mRNA DQ523561.1 - 20 - 21 1331 78

View Full Product Details