Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
shroom2
Entrez Gene ID
779568
Gene Name
shroom family member 2
Gene Aliases
apxl, shrm2
Chromosome Location
Chr.2: 72198081 - 72318414 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078710.1 NP_001072178.1 2 - 3 476 92
GenBank mRNA DQ886532.1 - 2 - 3 476 92

View Full Product Details