Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
shroom2
Entrez Gene ID
779568
Gene Name
shroom family member 2
Gene Aliases
apxl, shrm2
Chromosome Location
Chr.2: 72198081 - 72318414 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078710.1 NP_001072178.1 3 - 4 595 91
XM_012957215.2 XP_012812669.1 3 - 4 371 91
XM_018091332.1 XP_017946821.1 2 - 3 266 91
GenBank mRNA DQ886532.1 - 3 - 4 595 91

View Full Product Details