Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clba1
Entrez Gene ID
779636
Gene Name
clathrin binding box of aftiphilin containing 1
Gene Aliases
c14orf79
Chromosome Location
Chr.8: 79727637 - 79740974 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012968587.2 XP_012824041.1 5 - 6 1183 67
XM_012968588.2 XP_012824042.1 5 - 6 1000 67
GenBank mRNA BC118726.1 - 5 - 6 1000 67
CR761271.2 - 5 - 6 980 67

View Full Product Details