Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nsg2
Entrez Gene ID
779643
Gene Name
neuronal vesicle trafficking associated 2
Gene Aliases
caly3, hmp19
Chromosome Location
Chr.3: 33447786 - 33482202 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC118741.1 - 1 - 2 163 104
CU075860.1 - 1 - 2 166 104

View Full Product Details