Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rnf34
Entrez Gene ID
779671
Gene Name
ring finger protein 34, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.1: 153731109 - 153752689 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078756.1 NP_001072224.1 5 - 6 943 100
XM_012964975.2 XP_012820429.1 6 - 7 1107 100
XM_018094299.1 XP_017949788.1 5 - 6 1003 100
GenBank mRNA BC118805.1 - 5 - 6 943 100

View Full Product Details