Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
etaa1
Entrez Gene ID
779729
Gene Name
ETAA1, ATR kinase activator
Gene Aliases
-
Chromosome Location
Chr.5: 30499622 - 30509792 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078808.1 NP_001072276.1 1 - 2 109 85
GenBank mRNA BC121286.1 - 1 - 2 109 85

View Full Product Details