Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxo27
Entrez Gene ID
779735
Gene Name
F-box protein 27
Gene Aliases
-
Chromosome Location
Chr.8: 59651061 - 59667709 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078814.1 NP_001072282.1 2 - 3 528 101
XM_018096301.1 XP_017951790.1 2 - 3 389 101
GenBank mRNA BC121300.1 - 2 - 3 528 101
BC135462.1 - 2 - 3 468 101
CR762328.2 - 2 - 3 496 101

View Full Product Details