Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
txndc11
Entrez Gene ID
779736
Gene Name
thioredoxin domain containing 11
Gene Aliases
-
Chromosome Location
Chr.9: 31807457 - 31842149 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078815.1 NP_001072283.1 6 - 7 1837 74
GenBank mRNA BC121301.1 - 6 - 7 1837 74

View Full Product Details