Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
st6galnac4
Entrez Gene ID
779760
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
Gene Aliases
-
Chromosome Location
Chr.8: 2565944 - 2575958 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078839.1 NP_001072307.1 5 - 6 878 92
GenBank mRNA BC121364.1 - 5 - 6 878 92

View Full Product Details