Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
phyhipl
Entrez Gene ID
779774
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein-like
Gene Aliases
-
Chromosome Location
Chr.7: 7750854 - 7823637 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078853.1 NP_001072321.1 1 - 2 524 65
GenBank mRNA BC121389.1 - 1 - 2 524 65

View Full Product Details