Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
phyhipl
Entrez Gene ID
779774
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein-like
Gene Aliases
-
Chromosome Location
Chr.7: 7750854 - 7823637 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078853.1 NP_001072321.1 4 - 5 1023 65
XM_012966732.1 XP_012822186.1 4 - 5 611 65
XM_018095430.1 XP_017950919.1 4 - 5 597 65
GenBank mRNA BC121389.1 - 4 - 5 1023 65

View Full Product Details