Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl42
Entrez Gene ID
779854
Gene Name
mitochondrial ribosomal protein L42
Gene Aliases
-
Chromosome Location
Chr.3: 46674492 - 46679592 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078932.1 NP_001072400.1 4 - 5 325 125
GenBank mRNA BC121603.1 - 4 - 5 325 125
CT027939.1 - 4 - 5 411 125

View Full Product Details