Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ikbip
Entrez Gene ID
779867
Gene Name
IKBKB interacting protein
Gene Aliases
ikip
Chromosome Location
Chr.3: 12252948 - 12275293 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078945.1 NP_001072413.1 2 - 3 508 66
XM_018092167.1 XP_017947656.1 3 - 4 551 66
XM_018092168.1 XP_017947657.1 2 - 3 729 66
GenBank mRNA BC121637.1 - 2 - 3 508 66
BC135923.1 - 2 - 3 450 66
CR760287.2 - 2 - 3 545 66

View Full Product Details