Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lactb2
Entrez Gene ID
779879
Gene Name
lactamase beta 2
Gene Aliases
-
Chromosome Location
Chr.6: 105501391 - 105510931 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078957.1 NP_001072425.1 5 - 6 783 92
GenBank mRNA BC121666.1 - 5 - 6 783 92

View Full Product Details