Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
igbp1
Entrez Gene ID
779894
Gene Name
immunoglobulin (CD79A) binding protein 1
Gene Aliases
ibp1
Chromosome Location
Chr.8: 22752831 - 22760554 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078972.1 NP_001072440.1 4 - 5 790 81
GenBank mRNA BC121690.1 - 4 - 5 790 81

View Full Product Details