Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccdc85c
Entrez Gene ID
779899
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
Chromosome Location
Chr.8: 69634241 - 69727955 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078977.1 NP_001072445.1 2 - 3 1124 98
GenBank mRNA BC121701.1 - 2 - 3 1124 98
BC160578.1 - 2 - 3 1106 98

View Full Product Details