Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scrn2
Entrez Gene ID
779903
Gene Name
secernin 2
Gene Aliases
-
Chromosome Location
Chr.10: 4525469 - 4534541 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078981.1 NP_001072449.1 5 - 6 928 89
XM_012952558.2 XP_012808012.1 5 - 6 1037 89
XM_012952559.2 XP_012808013.1 5 - 6 1032 89
GenBank mRNA BC121706.1 - 5 - 6 928 89

View Full Product Details