Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dedd2
Entrez Gene ID
779936
Gene Name
death effector domain containing 2
Gene Aliases
-
Chromosome Location
Chr.7: 98942211 - 98962993 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079013.1 NP_001072481.1 5 - 6 1022 85
GenBank mRNA BC121906.1 - 5 - 6 1022 85

View Full Product Details