Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
isoc1
Entrez Gene ID
780052
Gene Name
isochorismatase domain containing 1
Gene Aliases
cgi-111
Chromosome Location
Chr.1: 158886624 - 158897260 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079129.1 NP_001072597.1 3 - 4 636 105
XM_012965593.2 XP_012821047.1 3 - 4 586 105
XM_012965599.2 XP_012821053.1 3 - 4 1125 105
XM_018094550.1 XP_017950039.1 3 - 4 506 105
GenBank mRNA BC122966.1 - 3 - 4 636 105

View Full Product Details