Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vgll4
Entrez Gene ID
780071
Gene Name
vestigial like family member 4
Gene Aliases
vgl-4
Chromosome Location
Chr.Un|NW_016683819: 56963 - 71795 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079147.1 NP_001072615.1 1 - 2 480 64
GenBank mRNA BC123005.1 - 1 - 2 480 64

View Full Product Details