Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cyb5d1
Entrez Gene ID
780076
Gene Name
cytochrome b5 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683788: 572 - 13102 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079152.1 NP_001072620.1 3 - 4 487 87
GenBank mRNA BC123012.1 - 3 - 4 487 87

View Full Product Details