Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppcs
Entrez Gene ID
780088
Gene Name
phosphopantothenoylcysteine synthetase
Gene Aliases
-
Chromosome Location
Chr.7: 77967650 - 77972119 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079164.1 NP_001072632.1 1 - 2 554 96
XM_012952575.2 XP_012808029.1 1 - 2 729 96
GenBank mRNA BC123078.1 - 1 - 2 554 96

View Full Product Details