Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
hmox2
Entrez Gene ID
780096
Gene Name
heme oxygenase 2
Gene Aliases
ho-2
Chromosome Location
Chr.Un|NW_016688369: 0 - 1203 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079172.1 NP_001072640.1 - 277 63
GenBank mRNA BC123915.1 - - 277 63

View Full Product Details