Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nadk2
Entrez Gene ID
780144
Gene Name
NAD kinase 2, mitochondrial
Gene Aliases
c5orf33, nadkd1
Chromosome Location
Chr.1: 183966358 - 183990568 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079219.1 NP_001072687.1 7 - 8 865 91
XM_012965806.2 XP_012821260.1 7 - 8 923 91
GenBank mRNA BC124020.1 - 7 - 8 865 91

View Full Product Details