Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ube2r2
Entrez Gene ID
780159
Gene Name
ubiquitin conjugating enzyme E2 R2
Gene Aliases
cdc34b, e2-cdc34b, ubc3b
Chromosome Location
Chr.1: 194653020 - 194678495 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079234.1 NP_001072702.1 2 - 3 434 92
GenBank mRNA BC124046.1 - 2 - 3 434 92

View Full Product Details