Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
htra1
Entrez Gene ID
780187
Gene Name
HtrA serine peptidase 1
Gene Aliases
Xhtra1, htra, l56, prss11
Chromosome Location
Chr.7: 10621200 - 10648559 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079262.2 NP_001072730.2 5 - 6 1367 66
GenBank mRNA BC125705.1 - 5 - 6 1367 66
BC135434.1 - 5 - 6 1290 66
CT025323.2 - 5 - 6 1274 66
CX740928.1 - - 543 66

View Full Product Details