Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
thap4
Entrez Gene ID
780282
Gene Name
THAP domain containing 4
Gene Aliases
-
Chromosome Location
Chr.5: 122184677 - 122196023 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079353.1 NP_001072821.1 4 - 5 1336 69
XM_012970575.2 XP_012826029.1 3 - 4 528 69
GenBank mRNA BC121669.1 - 4 - 5 1336 69

View Full Product Details