Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crls1
Entrez Gene ID
780303
Gene Name
cardiolipin synthase 1
Gene Aliases
-
Chromosome Location
Chr.5: 21070602 - 21077761 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079374.1 NP_001072842.1 5 - 6 797 86
GenBank mRNA BC121978.1 - 5 - 6 797 86

View Full Product Details