Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vps9d1
Entrez Gene ID
780322
Gene Name
VPS9 domain containing 1
Gene Aliases
c16orf7
Chromosome Location
Chr.4: 61711812 - 61735750 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079393.1 NP_001072861.1 4 - 5 591 104
GenBank mRNA BC123082.1 - 4 - 5 591 104
CR848484.2 - 4 - 5 628 104

View Full Product Details