Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egfl7
Entrez Gene ID
780371
Gene Name
EGF like domain multiple 7
Gene Aliases
xegfl7, zneu1
Chromosome Location
Chr.Un|NW_016683465: 73983 - 97575 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079441.1 NP_001072909.1 6 - 7 737 77
XM_012968703.1 XP_012824157.1 5 - 6 748 77
XM_012968704.2 XP_012824158.1 5 - 6 864 77
XM_018089875.1 XP_017945364.1 5 - 6 1044 77
GenBank mRNA BC123101.1 - 6 - 7 737 77

View Full Product Details